2-Thenoesäurehydrazid

Lieferant: MENAI

Synonyme: 2-Thiophencarbonsäurehydrazid, Thiophen-2-carbonsäurehydrazid, Thiophen-2-carbohydrazid

E2324.25 E2324.100
MENAE2324.25EA 0 EUR
MENAE2324.25 MENAE2324.100
2-Thenoesäurehydrazid
2-Thenoesäurehydrazid
Formel: C₅H₆N₂OS
Molecular Weight: 142,18 g/mol
Schmelzpunkt: 142…143 °C
MDL: MFCD00005435
CAS-Nummer: 2361-27-5
EINECS: 219-107-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR