1-(2,4-Dichlorphenyl)-2-thioharnstoff

Lieferant: MENAI

Synonyme: 2,4-Dichlorphenylthioharnstoff, N-(2,4-Dichlorphenyl)thioharnstoff

ER18.5 ER18.25 ER18.100
MENAER18.5EA 0 EUR
MENAER18.5 MENAER18.25 MENAER18.100
1-(2,4-Dichlorphenyl)-2-thioharnstoff
1-(2,4-Dichlorphenyl)-2-thioharnstoff
Formel: (C₆H₃Cl₂)NC(S)NH₂
Molecular Weight: 221.11 g/mol
Siedepunkt: 326
Schmelzpunkt: 160
MDL: MFCD00022161
CAS-Nummer: 6326-14-3
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR