N-(4-Bromphenyl)maleimid
Lieferant: MENAI
Synonyme:
N-(4-Bromphenyl)maleinsäureimid
IP18.50
IP18.100
MENAIP18.50EA
0
EUR
MENAIP18.50
MENAIP18.100
N-(4-Bromphenyl)maleimid
N-(4-Bromphenyl)maleimid
Formel:
C₁₀H₆BrNO₂ Molecular Weight: 252.07 g/mol Schmelzpunkt: 128.5…129.5 °C |
MDL:
MFCD00030664 CAS-Nummer: 13380-67-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...