(2,4-Dibromphenoxy)essigsäure

Lieferant: MENAI

IP35.25 IP35.100
MENAIP35.25EA 0 EUR
MENAIP35.25 MENAIP35.100
(2,4-Dibromphenoxy)essigsäure
(2,4-Dibromphenoxy)essigsäure
Formel: (C₆H₃Br₂)OCH₂COOH
Molecular Weight: 309,94 g/mol
MDL: MFCD00051478
CAS-Nummer: 10129-78-9
EINECS: 233-364-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR