(2,3-Xylyloxy)essigsäure

Lieferant: MENAI

Synonyme: (2,3-Dimethylphenoxy)essigsäure

IP37.50 IP37.100
MENAIP37.50EA 0 EUR
MENAIP37.50 MENAIP37.100
(2,3-Xylyloxy)essigsäure
(2,3-Xylyloxy)essigsäure
Formel: (C₆H₃(CH₃)₂)OCH₂COOH
Molecular Weight: 180.2 g/mol
MDL: MFCD00016814
CAS-Nummer: 2935-63-9
EINECS: 220-911-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR