(3,4-Xylyloxy)essigsäure

Lieferant: MENAI

IP38.50 IP38.100
MENAIP38.50EA 0 EUR
MENAIP38.50 MENAIP38.100
(3,4-Xylyloxy)essigsäure
(3,4-Xylyloxy)essigsäure
Formel: (C₆H₃(CH₃)₂)OCH₂COOH
Molecular Weight: 180.2 g/mol
MDL: MFCD00017595
CAS-Nummer: 13335-73-4
EINECS: 236-386-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR