3-Indolylessigsäurehydrazid

Lieferant: MENAI

Synonyme: Indol-3-ylessigsäurehydrazid, Indol-3-essigsäurehydrazid

NC2518.5 NC2518.25
MENANC2518.5EA 0 EUR
MENANC2518.5 MENANC2518.25
3-Indolylessigsäurehydrazid
3-Indolylessigsäurehydrazid
Formel: C₁₀H₁₁N₃O
Molecular Weight: 189.22 g/mol
Schmelzpunkt: 198
MDL: MFCD00005634
CAS-Nummer: 5448-47-5
EINECS: 226-672-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR