1-(2,6-Dichlorphenyl)-2-thioharnstoff
Lieferant: MENAI
|
Gefahr
|
Synonyme:
2,6-Dichlorphenylthioharnstoff, N-(2,6-Dichlorphenyl)thioharnstoff
SR464.5
SR464.25
SR464.100
MENASR464.5EA
0
EUR
MENASR464.5
MENASR464.25
MENASR464.100
1-(2,6-Dichlorphenyl)-2-thioharnstoff
1-(2,6-Dichlorphenyl)-2-thioharnstoff
Formel:
(C₆H₃Cl₂)NC(S)NH₂ Molecular Weight: 221.11 g/mol Siedepunkt: 326 Schmelzpunkt: 160…162 °C |
MDL:
MFCD00041156 CAS-Nummer: 6590-91-6 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...