1-(4-Methoxyphenyl)-2-thioharnstoff
Lieferant: MENAI
|
Gefahr
|
Synonyme:
4-Methoxyphenylthioharnstoff, N-(4-Methoxyphenyl)thioharnstoff
ST145.5
ST145.25
MENAST145.5EA
0
EUR
MENAST145.5
MENAST145.25
1-(4-Methoxyphenyl)-2-thioharnstoff
1-(4-Methoxyphenyl)-2-thioharnstoff
Formel:
(C₆H₄(OCH₃))NC(S)NH₂ Molecular Weight: 182.25 g/mol |
MDL:
MFCD00004936 CAS-Nummer: 2293-07-4 EINECS: 218-931-8 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...