1-Methoxy-5-methylphenaziniummethylsulfat

Lieferant: Serva

Synonyme: 1-Methoxy-N-methylphenazoniummethylsulfat

28966.01 28966.02
SERA28966.01EA 0 EUR
SERA28966.01 SERA28966.02
1-Methoxy-5-methylphenaziniummethylsulfat
1-Methoxy-5-methylphenazinium methyl sulphate
Formel: C₁₅H₁₆N₂O₅S
Molecular Weight: 336,37 g/mol
MDL: MFCD00040648
CAS-Nummer: 65162-13-2
EINECS: 265-579-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR