5-Methylphenaziniummethylsulfat
Lieferant: Serva
|
Gefahr
|
Synonyme:
PMS, N-Methylphenazoniummethylsulfat, Phenazinmethosulfat
32030.01
32030.02
SERA32030.01EA
38.1
EUR
SERA32030.01
SERA32030.02
5-Methylphenaziniummethylsulfat
5-Methylphenazinium methyl sulphate
Formel:
C₁₄H₁₄N₂O₄S Molecular Weight: 306,34 g/mol Schmelzpunkt: 198 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00011923 CAS-Nummer: 299-11-6 EINECS: 206-072-1 Merck Index: 12,06185 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...