Propidiumiodid

Lieferant: Serva
Achtung

Synonyme: 3,8-Diamino-5-(3-(diethylmethylammonio)propyl)-6-phenylphenanthridiniumdiiodid

33671.01 33671.02
SERA33671.01EA 148 EUR
SERA33671.01 SERA33671.02
Propidiumiodid
Propidiumiodid
Formel: C₂₇H₃₄I₂N₄
Molecular Weight: 668.4 g/mol
MDL: MFCD00011921
CAS-Nummer: 25535-16-4
EINECS: 247-081-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR