Amantadin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1-Aminotricyclo[3.3.1.1(3,7)]decan, 1-Adamantylamin, 1-Aminoadamantan
M-Clarity™ Qualitätsstufe = MQ200
138576-100G
138576-25G
138576-5G
SIAL138576-100GEA
495
EUR
SIAL138576-100G
SIAL138576-25G
SIAL138576-5G
Amantadin, Sigma-Aldrich®
Amantadin
Formel:
C₁₀H₁₇N Molecular Weight: 151.25 g/mol Siedepunkt: 214 Schmelzpunkt: 82 |
MDL:
MFCD00074732 CAS-Nummer: 768-94-5 EINECS: 212-201-2 Merck Index: 12,00389 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...