Ethyl-p-toluat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Methylbenzoesäureethylester, 4-Tolylsäureethylester, Ethyl-4-methylbenzoat, 4-Toluylsäureethylester, p-Toluylsäureethylester

M-Clarity™ Qualitätsstufe = MQ100

172693-250ML 172693-50ML
SIAL172693-250MLEA 106 EUR
SIAL172693-250ML SIAL172693-50ML
Ethyl-p-toluat, Sigma-Aldrich®
Ethyl-p-toluat
Formel: C₁₀H₁₂O₂
Molecular Weight: 164,2 g/mol
Dichte: 1,03 g/cm³ (20 °C)
MDL: MFCD00009117
CAS-Nummer: 94-08-6
EINECS: 202-301-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR