Anilin-[Ring-D5], Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Aniline-[ring-²H₅]
M-Clarity™ Qualitätsstufe = MQ200
175692-1G
175692-5G
SIAL175692-1GEA
143
EUR
SIAL175692-1G
SIAL175692-5G
Anilin-[Ring-D5], Sigma-Aldrich®
Anilin-[Ring-D5]
Formel:
C₆H₂D₅N Molecular Weight: 98,09 g/mol Schmelzpunkt: –6,3 °C Dichte: 1,077 g/cm³ (20 °C) |
CAS-Nummer:
4165-61-1 EINECS: 224-015-9 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...