1-Ethyl-3-methylimidazoliummethylsulfat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Ethyl-3-methyl-1H-imidazol-3-ium methyl sulphate

18086-50G-F 18086-5G-F
SIAL18086-50G-FEA 192 EUR
SIAL18086-50G-F SIAL18086-5G-F
1-Ethyl-3-methylimidazoliummethylsulfat, Sigma-Aldrich®
1-Ethyl-3-methylimidazolium methylsulphate
Formel: C₇H₁₄N₂O₄S
Molecular Weight: 222,27 g/mol
Dichte: 1,28 g/cm³ (20 °C)
MDL: MFCD08457697
CAS-Nummer: 516474-01-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR