1-Dodecyl-3-methylimidazoliumiodid, Sigma-Aldrich®

Lieferant: Merck

18289-50G-F 18289-5G-F
SIAL18289-50G-FEA 637 EUR
SIAL18289-50G-F SIAL18289-5G-F
1-Dodecyl-3-methylimidazoliumiodid, Sigma-Aldrich®
1-Dodecyl-3-methylimidazolium iodide
Formel: C₁₆H₃₁IN₂
Molecular Weight: 378,34 g/mol
CAS-Nummer: 81995-09-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR