2-(3-Methoxyphenyl)ethanol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-(3-Methoxyphenyl)ethylalkohol, 3-Methoxyphenethylalkohol

M-Clarity™ Qualitätsstufe = MQ100

187933-1G 187933-25G 187933-5G
SIAL187933-1GEA 27.2 EUR
SIAL187933-1G SIAL187933-25G SIAL187933-5G
2-(3-Methoxyphenyl)ethanol, Sigma-Aldrich®
2-(3-Methoxyphenyl)ethanol
Formel: C₉H₁₂O₂
Molecular Weight: 152,19 g/mol
MDL: MFCD00002893
CAS-Nummer: 5020-41-7
EINECS: 225-705-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR