2-Methylbenzofuran, Sigma-Aldrich®

Lieferant: Merck

224340-1G 224340-5G
SIAL224340-1GEA 166 EUR
SIAL224340-1G SIAL224340-5G
2-Methylbenzofuran, Sigma-Aldrich®
2-Methylbenzofuran
Formel: C₉H₈O
Molecular Weight: 132,16 g/mol
MDL: MFCD00005850
CAS-Nummer: 4265-25-2
EINECS: 224-249-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR