2-Benzylanilin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Aminodiphenylmethane, 2-Benzylaniline

M-Clarity™ Qualitätsstufe = MQ200

235350-10G 235350-50G
SIAL235350-10GEA 103 EUR
SIAL235350-10G SIAL235350-50G
2-Benzylanilin, Sigma-Aldrich®
2-Benzylanilin
Formel: C₁₃H₁₃N
Molecular Weight: 183,25 g/mol
MDL: MFCD00007750
CAS-Nummer: 28059-64-5
EINECS: 248-806-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR