3-Thenoesäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Thiophen-3-carbonsäure, 3-Thiophencarbonsäure
M-Clarity™ Qualitätsstufe = MQ100
247766-1G
247766-5G
SIAL247766-1GEA
31.8
EUR
SIAL247766-1G
SIAL247766-5G
3-Thenoesäure, Sigma-Aldrich®
3-Thenoesäure
Formel:
C₅H₄O₂S Molecular Weight: 128.15 g/mol Schmelzpunkt: 137 |
MDL:
MFCD00005467 CAS-Nummer: 88-13-1 EINECS: 201-802-5 Merck Index: 12,09415 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...