2,4,6-Triphenylanilin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5'-Phenyl-[1,1':3',1''-terphenyl]-2'-amine

M-Clarity™ Qualitätsstufe = MQ100

272515-1G 272515-5G
SIAL272515-1GEA 95 EUR
SIAL272515-1G SIAL272515-5G
2,4,6-Triphenylanilin, Sigma-Aldrich®
2,4,6-Triphenylanilin
Formel: C₂₄H₁₉N
Molecular Weight: 321,42 g/mol
MDL: MFCD00012371
CAS-Nummer: 6864-20-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR