N,N'-Diphenyl-p-phenylendiamin ≥98%, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
DPPD, N,N′-Diphenyl-1,4-phenylenediamine, N,N'-Diphenyl-1,4-phenylenediamine
M-Clarity™ Qualitätsstufe = MQ100
292265-25G
292265-5G
SIAL292265-25GEA
134
EUR
SIAL292265-25G
SIAL292265-5G
N,N'-Diphenyl-p-phenylendiamin ≥98%, Sigma-Aldrich®
N,N'-Diphenyl-p-phenylendiamin
Formel:
C₁₈H₁₆N₂ Molecular Weight: 260.34 g/mol Siedepunkt: 469 °C (1013 hPa) Schmelzpunkt: 146…149 °C Dichte: 1.21 g/cm³ (20 °C) |
MDL:
MFCD00003015 CAS-Nummer: 74-31-7 EINECS: 200-806-4 Merck Index: 12,03388 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...