2-Mercapto-1-methylimidazol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Mercapto-1-methyl-1H-imidazol, Thiamazol, 2-Mercapto-N-methyl-1H-imidazol, Methimazol, 2-Mercapto-N-methylimidazol
301507-25G
301507-5G
SIAL301507-25GEA
142
EUR
SIAL301507-25G
SIAL301507-5G
2-Mercapto-1-methylimidazol, Sigma-Aldrich®
2-Mercapto-1-methylimidazole
Formel:
C₄H₆N₂S Molecular Weight: 114,17 g/mol Siedepunkt: 155 °C (1013 hPa) Dichte: 1,36 g/cm³ (20 °C) |
MDL:
MFCD00179321 CAS-Nummer: 60-56-0 EINECS: 200-482-4 Merck Index: 13,06000 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...