4-Nitrophthalimid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4-Nitrophthalsäureimid, 4-Nitro-1,2-benzoldicarbonsäureimid
M-Clarity™ Qualitätsstufe = MQ100
332097-25G
332097-5G
SIAL332097-25GEA
430
EUR
SIAL332097-25G
SIAL332097-5G
4-Nitrophthalimid, Sigma-Aldrich®
4-Nitrophthalimid
Formel:
C₈H₄N₂O₄ Molecular Weight: 192,13 g/mol Schmelzpunkt: 215 °C |
MDL:
MFCD00005884 CAS-Nummer: 89-40-7 EINECS: 201-905-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...