3-(3-Methoxyphenyl)propionsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3-(3-Methoxyphenyl)propansäure
M-Clarity™ Qualitätsstufe = MQ100
349763-25G
349763-5G
SIAL349763-25GEA
159
EUR
SIAL349763-25G
SIAL349763-5G
3-(3-Methoxyphenyl)propionsäure, Sigma-Aldrich®
3-(3-Methoxyphenyl)propionsäure
Formel:
C₁₀H₁₂O₃ Molecular Weight: 180,2 g/mol |
MDL:
MFCD00014027 CAS-Nummer: 10516-71-9 EINECS: 234-049-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...