1-Acetylindol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-Acetylindol, 1-Indolylmethylketon, 1-Indolyl methyl ketone, 1-(1-Indolyl)-1-ethanon, Indol-1-ylmethylketon

377104-1G 377104-5G
SIAL377104-1GEA 103 EUR
SIAL377104-1G SIAL377104-5G
1-Acetylindol, Sigma-Aldrich®
1-Acetylindol
Formel: C₁₀H₉NO
Molecular Weight: 159,19 g/mol
Siedepunkt: 97,5…101 °C (0.1 mmHg)
Dichte: 1,387 g/cm³ (20 °C)
MDL: MFCD00038005
CAS-Nummer: 576-15-8
EINECS: 209-396-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR