Tris(dimethylamino)phosphin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Hexamethylphosphortriamid
M-Clarity™ Qualitätsstufe = MQ100
393290-25ML
393290-5ML
SIAL393290-25MLEA
272
EUR
SIAL393290-25ML
SIAL393290-5ML
Tris(dimethylamino)phosphin, Sigma-Aldrich®
Tris(dimethylamino)phosphin
Formel:
C₆H₁₈N₃P Molecular Weight: 163.2 g/mol Siedepunkt: 150 Schmelzpunkt: –44 °C Dichte: 0.8962 g/cm³ (20 °C) |
MDL:
MFCD00008301 CAS-Nummer: 1608-26-0 EINECS: 216-534-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...