2-(3-Bromphenyl)ethanol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

394521-1G 394521-5G
SIAL394521-1GEA 26 EUR
SIAL394521-1G SIAL394521-5G
2-(3-Bromphenyl)ethanol, Sigma-Aldrich®
2-(3-Bromphenyl)ethanol
Formel: C₈H₉BrO
Molecular Weight: 201,06 g/mol
MDL: MFCD00191857
CAS-Nummer: 28229-69-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR