1-Benzyl-3-methylimidazoliumtetrafluoroborat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Benzyl-3-methyl-1H-imidazol-3-ium tetrafluoroborate

40819-5G-F 40819-50G-F
SIAL40819-5G-FEA 51 EUR
SIAL40819-5G-F SIAL40819-50G-F
1-Benzyl-3-methylimidazoliumtetrafluoroborat, Sigma-Aldrich®
1-Benzyl-3-methylimidazoliumtetrafluoroborat
Formel: C₁₁H₁₃BF₄N₂
Molecular Weight: 260,04 g/mol
MDL: MFCD07784449
CAS-Nummer: 500996-04-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR