3-Aminoisoxazol, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
424218-25ML
424218-5ML
SIAL424218-25MLEA
444
EUR
SIAL424218-25ML
SIAL424218-5ML
3-Aminoisoxazol, Sigma-Aldrich®
3-Aminoisoxazole
Formel:
C₃H₄N₂O Molecular Weight: 84,08 g/mol |
MDL:
MFCD00038814 CAS-Nummer: 1750-42-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...