2,4-Dimethoxybenzylamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(2,4-Dimethoxyphenyl)methanamine, (2,4-Dimethoxyphenyl)methanamine

M-Clarity™ Qualitätsstufe = MQ100

432725-25ML 432725-5ML
SIAL432725-25MLEA 211 EUR
SIAL432725-25ML SIAL432725-5ML
2,4-Dimethoxybenzylamin, Sigma-Aldrich®
2,4-Dimethoxybenzylamin
Formel: C₉H₁₃NO₂
Molecular Weight: 167,21 g/mol
MDL: MFCD00052393
CAS-Nummer: 20781-20-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR