Tetramethylammoniumfluorid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
459135-1G
459135-5G
SIAL459135-1GEA
110
EUR
SIAL459135-1G
SIAL459135-5G
Tetramethylammoniumfluorid, Sigma-Aldrich®
Tetramethylammonium fluoride
Formel:
C₄H₁₂FN Molecular Weight: 93,14 g/mol Schmelzpunkt: 319 °C Dichte: 1,058 g/cm³ (20 °C) |
CAS-Nummer:
373-68-2 EINECS: 206-769-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...