1-Ethyl-3-methylimidazoliumhexafluorophosphat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Ethyl-3-methyl-1H-imidazol-3-ium hexafluorophosphate(V)

46093-50G-F 46093-5G-F
SIAL46093-50G-FEA 1050 EUR
SIAL46093-50G-F SIAL46093-5G-F
1-Ethyl-3-methylimidazoliumhexafluorophosphat, Sigma-Aldrich®
1-Ethyl-3-methylimidazoliumhexafluorophosphate
Formel: C₆H₁₁F₆N₂P
Molecular Weight: 256,13 g/mol
MDL: MFCD00216703
CAS-Nummer: 155371-19-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR