5-Brom-2-thenoesäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
5-Bromthiophen-2-carbonsäure, 5-Brom-2-thiophencarbonsäure
M-Clarity™ Qualitätsstufe = MQ200
467944-1G
467944-5G
SIAL467944-1GEA
26
EUR
SIAL467944-1G
SIAL467944-5G
5-Brom-2-thenoesäure, Sigma-Aldrich®
5-Brom-2-thenoesäure
Formel:
C₅H₃BrO₂S Molecular Weight: 207,05 g/mol |
MDL:
MFCD00079725 CAS-Nummer: 7311-63-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...