1-Iod-3,3,3-trifluorpropan, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1,1,1-Trifluor-3-iodpropan
M-Clarity™ Qualitätsstufe = MQ100
473820-1G
473820-5G
SIAL473820-1GEA
22.8
EUR
SIAL473820-1G
SIAL473820-5G
1-Iod-3,3,3-trifluorpropan, Sigma-Aldrich®
1-Iod-3,3,3-trifluorpropan
Formel:
C₃H₄F₃I Molecular Weight: 223,96 g/mol |
MDL:
MFCD00038531 CAS-Nummer: 460-37-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...