(Trifluormethyl)trimethylsilan, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
trimethyl(trifluoromethyl)silane
M-Clarity™ Qualitätsstufe = MQ200
488712-25ML
488712-5ML
SIAL488712-25MLEA
344
EUR
SIAL488712-25ML
SIAL488712-5ML
(Trifluormethyl)trimethylsilan, Sigma-Aldrich®
(Trifluoromethyl)trimethylsilane
Formel:
C₄H₉F₃Si Molecular Weight: 142,2 g/mol Siedepunkt: 55,5 °C (1013 hPa) Dichte: 0,95 g/cm³ (20 °C) |
MDL:
MFCD00145454 CAS-Nummer: 81290-20-2 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...