1-Benzyl-3-methylimidazoliumchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Benzyl-3-methyl-1H-imidazol-3-ium chloride

49914-5G-F 49914-50G-F
SIAL49914-5G-FEA 56.1 EUR
SIAL49914-5G-F SIAL49914-50G-F
1-Benzyl-3-methylimidazoliumchlorid, Sigma-Aldrich®
1-Benzyl-3-methylimidazoliumchlorid
Formel: C₁₁H₁₃ClN₂
Molecular Weight: 208,69 g/mol
MDL: MFCD07784451
CAS-Nummer: 36443-80-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR