6-Fluor-m-anisaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Fluor-5-methoxybenzaldehyd, 6-Fluor-3-anisaldehyd

M-Clarity™ Qualitätsstufe = MQ100

542644-1G-A 542644-5G-A
SIAL542644-1G-AEA 35 EUR
SIAL542644-1G-A SIAL542644-5G-A
6-Fluor-m-anisaldehyd, Sigma-Aldrich®
6-Fluor-m-anisaldehyd
Formel: C₈H₇FO₂
Molecular Weight: 154,14 g/mol
MDL: MFCD00070795
CAS-Nummer: 105728-90-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR