2-Chlorisonicotinonitril, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Chlor-4-cyanpyridin, 2-Chlor-4-pyridincarbonitril, 2-Chlor-4-pyridincarbonsäurenitril
M-Clarity™ Qualitätsstufe = MQ100
548227-25G
548227-5G
SIAL548227-25GEA
120
EUR
SIAL548227-25G
SIAL548227-5G
2-Chlorisonicotinonitril, Sigma-Aldrich®
2-Chlorisonicotinonitril
Formel:
C₆H₃ClN₂ Molecular Weight: 138,56 g/mol |
MDL:
MFCD00174318 CAS-Nummer: 33252-30-1 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...