3,5-Dimethylbenzolsulfonylchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: m-Xylol-5-sulfonsäurechlorid, m-Xylol-5-sulfonylchlorid, 3,5-Dimethylbenzol-1-sulfonylchlorid, 3,5-Dimethylbenzolsulfonsäurechlorid, 3,5-Dimethylbenzolsulfochlorid

M-Clarity™ Qualitätsstufe = MQ200

552240-1G 552240-5G
SIAL552240-1GEA 42.1 EUR
SIAL552240-1G SIAL552240-5G
3,5-Dimethylbenzolsulfonylchlorid, Sigma-Aldrich®
3,5-Dimethylbenzolsulfonylchlorid
Formel: C₈H₉ClO₂S
Molecular Weight: 204,68 g/mol
MDL: MFCD03094655
CAS-Nummer: 2905-27-3
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR