3-(4-Aminophenyl)propionsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-(4-Aminophenyl)propansäure

M-Clarity™ Qualitätsstufe = MQ100

560251-1G 560251-5G
SIAL560251-1GEA 30.6 EUR
SIAL560251-1G SIAL560251-5G
3-(4-Aminophenyl)propionsäure, Sigma-Aldrich®
3-(4-Aminophenyl)propionsäure
Formel: C₉H₁₁NO₂
MDL: MFCD00017118
CAS-Nummer: 2393-17-1
EINECS: 219-245-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR