6-Brom-o-toluidin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

563048-5G 563048-1G
SIAL563048-5GEA 225 EUR
SIAL563048-5G SIAL563048-1G
6-Brom-o-toluidin, Sigma-Aldrich®
6-Brom-o-toluidin
Formel: C₇H₈BrN
Molecular Weight: 186,05 g/mol
MDL: MFCD04039885
CAS-Nummer: 53848-17-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR