2-Iod-4-nitroanilin, Sigma-Aldrich®

Lieferant: Merck

576689-1G 576689-5G
SIAL576689-1GEA 26 EUR
SIAL576689-1G SIAL576689-5G
2-Iod-4-nitroanilin, Sigma-Aldrich®
2-Iod-4-nitroanilin
Formel: C₆H₅IN₂O₂
Molecular Weight: 264,02 g/mol
MDL: MFCD00955758
CAS-Nummer: 6293-83-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR