(3-Phenoxyphenyl)essigsäure, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
3-Phenoxyphenylacetic acid
M-Clarity™ Qualitätsstufe = MQ100
590827-1G
590827-5G
SIAL590827-1GEA
54.6
EUR
SIAL590827-1G
SIAL590827-5G
(3-Phenoxyphenyl)essigsäure, Sigma-Aldrich®
(3-Phenoxyphenyl)essigsäure
Formel:
(C₆H₅OC₆H₅)CH₂COOH Molecular Weight: 228,25 g/mol |
MDL:
MFCD00016826 CAS-Nummer: 32852-81-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...