2-Brom-4-fluor-1-iodbenzol, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 1-Brom-5-fluor-2-iodbenzol, 3-Brom-1-fluor-4-iodbenzol

M-Clarity™ Qualitätsstufe = MQ100

594326-1G 594326-5G
SIAL594326-1GEA 50.6 EUR
SIAL594326-1G SIAL594326-5G
2-Brom-4-fluor-1-iodbenzol, Sigma-Aldrich®
2-Brom-4-fluor-1-iodbenzol
Formel: C₆H₃BrFI
Molecular Weight: 300,9 g/mol
MDL: MFCD00142868
CAS-Nummer: 202865-73-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR