Kaliumtellurit, Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyme: di-Kaliumtellurtrioxid, Kaliumtellur(IV)oxid

60539-10G 60539-50G
SIAL60539-10GEA 54.2 EUR
SIAL60539-10G SIAL60539-50G
Kaliumtellurit, Sigma-Aldrich®
Kaliumtellurit
Formel: K₂TeO₃
Molecular Weight: 253,79 g/mol
MDL: MFCD00011379
CAS-Nummer: 7790-58-1
EINECS: 232-213-1
UN: 3284
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR