2-Brom-N-methylanilin, Sigma-Aldrich®

Lieferant: Merck

630918-1G 630918-5G
SIAL630918-1GEA 100 EUR
SIAL630918-1G SIAL630918-5G
2-Brom-N-methylanilin, Sigma-Aldrich®
2-Brom-N-methylanilin
Formel: C₇H₈BrN
Molecular Weight: 186,05 g/mol
MDL: MFCD06798064
CAS-Nummer: 6832-87-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR