3-Brom-N-methylanilin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

630985-1G 630985-5G
SIAL630985-1GEA 32.3 EUR
SIAL630985-1G SIAL630985-5G
3-Brom-N-methylanilin, Sigma-Aldrich®
3-Brom-N-methylanilin
Formel: C₇H₈BrN
Molecular Weight: 186,05 g/mol
MDL: MFCD05664376
CAS-Nummer: 66584-32-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR