7-Indolcarbaldehyd, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Indol-7-carboxaldehyd
M-Clarity™ Qualitätsstufe = MQ100
632414-1G
632414-5G
SIAL632414-1GEA
59.2
EUR
SIAL632414-1G
SIAL632414-5G
7-Indolcarbaldehyd, Sigma-Aldrich®
7-Indolcarbaldehyd
Formel:
C₉H₇NO Molecular Weight: 145,16 g/mol |
MDL:
MFCD01318152 CAS-Nummer: 1074-88-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...